Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Aeroplysinin, Antiangiogenic Agent, >95% Purity, 1 mg

Catalog No. 502706210
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6210 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6210 Supplier Abcam Supplier No. AB1426851MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent antiangiogenic agent that inhibits COX-2 expression and induces apoptosis in endothelial cells.

  • Molecular Weight: 338.98 Da
  • Purity: >95%
  • Potent antiangiogenic agent: Selectivity for endothelial cells; inhibits COX-2 expression
  • Biological activity: Induces apoptosis via caspase activation, chromatin condensation, and nuclear fragmentation
  • Other effects: Antibiotic and cytotoxic (IC50 = 3 μM); displays wide-spectrum antitumor activity; active in vivo
  • Chemical Name: 2-[(1S,6R)-3,5-dibromo-1,6-dihydroxy-4-methoxycyclohexa-2,4-dien-1-yl]acetonitrile
  • CAS Number: 28656-91-9

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 95% 2-[(1S,6R)-3,5-dibromo-1,6-dihydroxy-4-methoxycyclohexa-2,4-dien-1-yl]acetonitrile
Content And Storage Store at -20°C.
Shipping Condition Ambient
Target Molecule COX-2
Formulation 95% 2-[(1S,6R)-3,5-dibromo-1,6-dihydroxy-4-methoxycyclohexa-2,4-dien-1-yl]acetonitrile
Quantity 1 mg
For Use With (Application) Antiangiogenesis, apoptosis induction, antitumor research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.