Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Aeroplysinin, Antiangiogenic Agent, 95% Purity, 100 μg

Catalog No. 502706211
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6211 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6211 Supplier Abcam Supplier No. AB142685100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent antiangiogenic agent that inhibits COX-2 expression and induces apoptosis.

  • Molecular Weight: 338.98 Da
  • Purity: >95%
  • Potent antiangiogenic agent that inhibits COX-2 expression
  • Selectivity: Endothelial cells
  • Mechanism of Action: Induces apoptosis via caspase activation, chromatin condensation and nuclear fragmentation
  • Biological Activity: Antibiotic and cytotoxic effects (IC50 = 3 μM) with wide-spectrum antitumor activity
  • In vivo: Active
  • Chemical Name: 2-[(1S,6R)-3,5-dibromo-1,6-dihydroxy-4-methoxycyclohexa-2,4-dien-1-yl]acetonitrile

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule COX-2
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Buffer 95% 2-[(1S,6R)-3,5-dibromo-1,6-dihydroxy-4-methoxycyclohexa-2,4-dien-1-yl]acetonitrile
Content And Storage Store at -20°C.
Quantity 100 μg
For Use With (Application) Antiangiogenesis, apoptosis induction, tumor research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.