Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Aerosol Filter Tips, Non-sterile

Catalog No. 14222764 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
0.5 to 20 μL
1 to 200 μL
100 to 1000 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-222-764 100 to 1000 μL
14-222-775 0.5 to 20 μL
14-222-776 1 to 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-222-764 Supplier Axygen™ Supplier No. TF1000
Add to Cart
Add to Cart

Prevent cross-contamination of samples. Axygen™ Aerosol Filter Tips, Non-sterile feature a universal fit. All tips comply with USP 3127 (Heavy Metal Contamination) and are free of leachable additives. Compatible with a variety of pipettes and available in a range of filter sizes.

  • Technologically advanced filter material allows free passage of air while blocking aerosol contamination.
  • Filter membrane contains no leachable additives.
  • Axygen™ Maxymum Recovery™ surface improves accuracy and ensures maximum sample recovery.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
  • Order Info

    Available in bulk or racked formats.

    Certifications

    Certified RNase-, DNase- and Pyrogen free

    TRUSTED_SUSTAINABILITY

    Specifications

    Volume (Metric) 100 to 1000 μL
    Volume (Metric) Dispensing 100 to 1000 μL
    Filtered Filtered
    Sterility Non-sterile
    Color Clear
    Autoclavable Not Autoclavable
    Certifications/Compliance USP 3127
    Clean Claims DNase, RNase, and Pyrogen Free
    For Use With (Application) DNA Amplification, Microbiological Applications
    Format Bulk
    Tip Style Universal
    Packaging 1,000 Tips/Pk., 5 Packs/Cs.
    Quantity 1000/Pk.
    Product Type Aerosol Barrier Tip
    Green Features Bulk Handheld Tips Use on Average 74% Less Resin Than Their Pre-racked Alternatives.
    For Use With (Equipment) Universal Fit
    Material Polypropylene
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.