Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Aerosol Filter Tips, Sterile
GSA_VA

Catalog No. 14222777 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
0.5 to 20 μL
0.5 to 50 μL
1 to 100 μL
1 to 200 μL
1 to 300 μL
100 to 1000 μL
100 to 1100 μL
15 to 1250 μL
Tip Style:
Beveled
Matrix-style
Maxymum Recovery
Maxymum Recovery, Ultra-micro
Ultra-micro
Universal
Format:
Bulk
Rack
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style Format
14-222-777 1 to 200 μL Maxymum Recovery Rack
14-222-219 15 to 1250 μL Matrix-style Rack
14-222-763 1 to 100 μL Universal Bulk
14-222-765 100 to 1000 μL Maxymum Recovery Rack
14-222-770 1 to 100 μL Universal Rack
14-222-766 100 to 1000 μL Universal Rack
14-222-769 1 to 100 μL Maxymum Recovery Rack
14-222-772 100 to 1100 μL Beveled Rack
14-222-778 1 to 200 μL Universal Rack
14-222-781 0.5 to 20 μL Maxymum Recovery Rack
14-222-782 0.5 to 20 μL Universal Rack
14-222-787 1 to 300 μL Maxymum Recovery Rack
14-222-788 1 to 300 μL Universal Rack
14-222-794 0.5 to 20 μL Maxymum Recovery, Ultra-micro Rack
14-222-795 0.5 to 20 μL Ultra-micro Rack
14-222-799 0.5 to 50 μL Maxymum Recovery Rack
14-222-800 0.5 to 50 μL Universal Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 14-222-777 Supplier Axygen™ Supplier No. TF200LRS
Add to Cart
Add to Cart

Prevent cross-contamination with robotic barrier tips. Axygen™ Aerosol Filter Tips, Sterile are e-beam irradiated to prevent product degradation and are free of leachable additives. Available in a wide variety of tip styles.

  • Technologically advanced filter barrier pipette tips
  • Ideal for DNA amplification and microbiological applications where trace contamination can invalidate results
  • Filter allows free passage of air while preventing cross-contamination by blocking the passage of aerosols or liquids
  • Nonpyrogenic, clear polyethylene filters are free from DNA inhibitors and leachable additives
  • Every filter is inserted into the tip robotically to ensure freedom from nuclease contamination
  • Packaged in sealed-bottom racks with lids to prevent contamination
  • Wide variety of tip styles available in a range of packaging quantities:
    • Maxymum Recovery™ technology provides excellent accuracy and prevents wastage of precious samples
    • Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures

Order Info

Available in bulk or racked formats.

Certifications

Certified RNase-, DNase- and Pyrogen free

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Volume (Metric) Dispensing 1 to 200 μL
Filtered Filtered
Sterility Sterile
Color Clear
Rack Color Clear
Autoclavable Autoclavable
Certifications/Compliance ISO 13485
Clean Claims DNase, RNase, and Pyrogen Free
For Use With (Application) DNA Amplification, Microbiological Applications
Format Rack
Tip Style Maxymum Recovery
Packaging Rack
Quantity 960/Pk.
Product Type Universal Fit Filter Pipet Tip
Includes 12 Racks x 96 Tips
Green Features No Intentionally Added PFAS
ACT Label No
For Use With (Equipment) Gilson Pipetman P200
Graduated Yes
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.