Learn More
Thermo Scientific™ AerosolSense™ Cartridges
Catalog No. 12148001 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
AerosolSense Sampler
Catalog No. For Use With (Equipment)
12-148-001 AerosolSense Sampler
1 options
Catalog No. 12-148-001 Supplier Thermo Scientific™ Supplier No. 12148001
Used with the Thermo Scientific™ AerosolSense™ Sampler
  • Small and easy to use
  • Isolated collection substrate
  • Can only be used with the Thermo Scientific™ AerosolSense™ Sampler
TRUSTED_SUSTAINABILITY
Specifications
Product Type AerosolSense™ Cartridges
For Use With (Equipment) AerosolSense Sampler
Quantity 30 Pk.