Learn More
Medchemexpress LLC AES-135 100mg | 2361659-61-0 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY114483100MG
Encompass_Preferred
AES-135 is a hydroxamic acid-based pan-histone deacetylase (HDAC) inhibitor for research use. It inhibits HDAC3, HDAC6, HDAC8, and HDAC11 with reported IC50s of approximately 190-1,100 nM and has shown activity in an orthotopic pancreatic cancer mouse model.
  • Pan-HDAC inhibitor with reported IC50s of ~190-1,100 nM.
  • High reported purity: 98.2%.
  • Available as solid or as 10 mM solution in DMSO formats.
  • Offered in common research pack sizes, including 100 mg.
  • Intended for in vitro and in vivo preclinical research use only.
Catalog No. 50-002-29031