Learn More
Medchemexpress LLC AES-135 10mg | 2361659-61-0 | 10 MG

Supplier: Medchemexpress LLC HY11448310MG
AES-135 is a hydroxamic acid-based pan-histone deacetylase (HDAC) inhibitor for research. It inhibits HDAC3, HDAC6, HDAC8, and HDAC11 (reported IC50 range 190-1,100 nM) and has been used in preclinical cancer models.
- Inhibits HDAC3, HDAC6, HDAC8, and HDAC11; IC50 ~190-1,100 nM.
- Chemical formula C33H29F6N3O5S; molecular weight 693.66 g·mol⁻¹.
- Purity typically ≥98% by HPLC.
- Supplied as a 10 mg powder; store powder at -20°C for long-term stability.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.