Learn More
Medchemexpress LLC AES-135 25mg | 2361659-61-0 | 25 MG

Supplier: Medchemexpress LLC HY11448325MG
AES-135 is a hydroxamic acid-based histone deacetylase (HDAC) inhibitor active against HDAC3, HDAC6, HDAC8, and HDAC11. It is provided as a research reagent in solid and solution formats for preclinical biochemical and cell-based studies.
- Hydroxamic acid-based HDAC inhibitor active against HDAC3, HDAC6, HDAC8, and HDAC11
- Purity 98.2%
- Molecular weight 693.66 g/mol
- Chemical formula C33H29F6N3O5S
- Available as solid and as 10 mM solution in DMSO
- Intended for biochemical and cell-based research applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.