Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AF9 (MLLT3) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$207.00 - $524.00
Specifications
| Antigen | AF9 (MLLT3) |
|---|---|
| Concentration | 1.0 mg/mL |
| Applications | ChIP sequencing (ChIP-seq), Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ABE259725UG
|
MilliporeSigma
ABE259725UG |
25 μg |
Each for $207.00
|
|
|||||
|
ABE2597100
|
MilliporeSigma
ABE2597100UG |
100 μg |
Each for $524.00
|
|
|||||
Description
AF9 (MLLT3) Polyclonal antibody specifically detects AF9 (MLLT3) in Human, Mouse samples. It is validated for ChIP-Seq, Immunocytochemistry, Western Blotting.Specifications
| AF9 (MLLT3) | |
| ChIP sequencing (ChIP-seq), Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal region of human AF9 (MLLT3) protein. | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. |
| 1.0 mg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title