Learn More
Market Lab AFB Control Slides, ML62220, 10pk
Supplier:  Market Lab ML62220
Encompass_Preferred
The AFB Control Slides ML62220 are a set of 12 slides used for quality control in testing for acid-fast bacteria. They contain smears of Mycobacterium tuberculosis, Mycobacterium kansasii, Mycobacterium scrofulaceum, and Mycobacterium avium. These slides are intended for laboratory use with fluorescence microscopy for acid-fast staining. They allow laboratories to monitor test sensitivity and staining quality. The slides are permanently stained, ready-to-use, and come in a cardboard storage box.
Catalog No. NC2580627
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.