Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Afinion™ ACR Test Kit Rx

Catalog No. 23050102 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
15/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-050-102 15/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-050-102 Supplier Abbott™ Supplier No. 1116972
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A test for quantitative determination of albumin, creatinine, and albumin/creatinine ratio (ACR) in human urine. Used for early identification of renal disease in patients with diabetes and/or hypertension.

  • 3.5 μL random/spot urine sample
  • Higly accurate results in 5 minutes
  • Increased accuracy by measuring creatinine
  • Quality control and operator lockout
  • No sample preparation
  • Measuring Range
    • Albumin: 5.0 - 200.0 mg/L
    • Creatinine: 16.4 - 339.9 mg/dL (1.5 - 30.0 mmol/L)
    • ACR: 1.0 - 1225.0 mg/g (0.1 - 140.0 mg/mmol)

Specifications

Description NGSP and IFCC Certified
Clia Complexity Moderate
Sample Type Human Urine
For Use With (Application) Quantitative Determination of Albumin, Creatinine, and Albumin/Creatinine Ratio (ACR)
Quantity 15/Pk.
Range Albumin: 5.0 to 200.0 mg/L, Creatinine: 16.4 to 339.9 mg/dL (1.5 to 30.0 mmol/L), ACR: 1.0 to 1225.0 mg/g (0.1 to 140.0 mg/mmol)
Sample Size 3.5 μL
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.