Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Afinion™ HbA1c Test Kit Rx

Catalog No. 23050105 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
15/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-050-105 15/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-050-105 Supplier Abbott™ Supplier No. 1116974
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Afinion™ HbA1c Assay is a test for quantitative determination of glycated hemoglobin (HbA1c) in human whole blood, used to monitor metabolic control in patients with diabetes. Immediate point of care HbA1c results for efficient monitoring of glycemic control.

  • Capillary and venous whole blood sample material
  • 1.5 μL sample volume
  • Measuring range: 4% to 15% HbA1c
  • Storage: 3 months at room temperature
  • Ready-to-use controls in two levels

Specifications

Content And Storage Up to 3 months (at Room Temperature)
Sample Type Capillary Blood or Anticoagulated Venous Blood (EDTA, Citrate or Heparin)
For Use With (Application) Quantitative Determination of Glycated Hemoglobin (HbA1c)
Methods Compatibility Traceable to the IFCC Reference Method
Quantity 15/Pk.
Range 4% to 15% HbA1c (20 to 140 mmol/mol)
Sample Size 1.5 μL
Test Time 3 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.