Learn More
Cayman Chemical Aflatoxin B1, 1162-65-8, 5 mg
SDP
Supplier:  Cayman Chemical 112935
Encompass_Preferred
Molecular Formula C17H12O6, Purity ≥98%, Formula Weight 312.3, Synonyms: 2,3,6aR,9aS-tetrahydro-4-methoxy-1H,11H-cyclopenta[c]furo[3',2':4,5]furo[2,3-h][1]benzopyran-1,11-dione, AFB1, HSDB 3453, NSC 529592; Naturally occuring mycotoxin and food contaminant that induces the transversion of G to T at codon 249 of the p53 tumor suppressor gene and is a likely pathogen that causes hepatocellular carcinoma.
Catalog No. NC1026133