Learn More
Abcam Aflatoxin B1 (AFB1) ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB3029331X96TESTS
Encompass_Preferred
Aflatoxin B1 (AFB1) ELISA Kit is a Competitive ELISA kit for the measurement of Aflatoxin B1 (AFB1) in Food samples samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-0738
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.