Learn More
Enzo Life Sciences Aflatoxin M2 (0.1mg). CAS: 6885-57-0
SDP
Supplier:  Enzo Life Sciences ALX630114MC01
Encompass_Preferred
Potent hepatotoxic and hepatocarcinogenic mycotoxin. Metabolite of aflatoxin B2. DNA-damaging agent. Alternative name: 4-Hydroxyaflatoxin B2. Purity: ≥98% (HPLC, TLC). Appearance: Off-white powder. Blue-violet fluorescence. Solubility: Soluble in DMSO or methanol; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9640
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.