Learn More
Description
This antibody reacts with human alpha-Fetoprotein (AFP), a 70 kDa oncofetal antigen. AFP is a major fetal plasma protein, but is not present in healthy adult tissues. Elevated AFP concentrations in adult plasma may be an early marker of hepatocellular carcinoma or teratoblastoma, while high concentrations in amniotic fluid may indicate severe congenital defects of a fetus. The MA1-19342 anti-alpha Fetoprotein antibody (clone AFP-11) has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody MA1-19178 (clone AFP-01). Immunohistochemistry (Paraffin): Heat mediated antigen retrieval (sodium citrate buffer); mAb incubation 1 hour / RT, detection DAB; Western Blot: non-reducing conditions.
Specifications
Specifications
| Antigen | AFP |
| Applications | ELISA, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | AFP-11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | AFP |
| Gene Accession No. | P02771 |
| Gene Alias | a fetoglobulin; a fetoprotein; Afp; AFPD; Alpha fetoglobulin; alpha fetoprotein; alpha-1-fetoprotein; Alpha-fetoglobulin; alpha-fetoprotein; alpha-fetoprotein precursor; alpha-foetoprotein; chloride intracellular channel 6; FETA; fetoglobulin; fetoprotein; HPAFP; a fetoglobulin; a fetoprotein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
