Learn More
Medchemexpress LLC HY-N2991 1mg Medchemexpress, Dehydropachymic acid CAS:77012-31-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN29911MG
Encompass_Preferred
Medchemexpress, HY-N2991 1mg Dehydropachymic acid CAS:77012-31-8 Dehydropachymic acid is one of the major triterpenes isolated from Poria cocos. Dehydropachymic acid is more effective in autophagy-lysosome pathway (ALP) impaired cells rather than normal cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2911
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.