Learn More
Enzo Life Sciences AG-490 (10mg). CAS: 133550-30-8
SDP
Supplier:  Enzo Life Sciences BMLEI2720010
Encompass_Preferred
AG-490 is a potent inhibitor of the JAK-2 tyrosine kinase. In acute lymphoblastic leukemia (ALL) cells, which abundantly express JAK-2, AG-490 dose-dependently inhibited DNA synthesis, blocked cell growth and induced apoptosis. At 5 µM, AG-490 almost completely blocked the growth of all pre-B ALL cells but had no significant effect on the growth of mitogen-stimulated normal B or T cells, B-cell lymphoma or T-cell leukemia cells. AG-490 does not significantly inhibit other kinases such as Lck, Lyn, Btk, Syk and Src. It blocks interleukin-7-induced JAK kinase activity in T-cells (JAK-1, JAK-3) and the consequent phosphorylation of PI-3 kinase. AG-490 is cell permeable and is a valuable tool for studying the cellular role of JAK kinases in signal transduction. Alternative name: N-Benzyl-3,4-dihydroxybenzylidenecyanoacetamide. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (30mg/ml), dimethyl formamide (40mg/ml) or 100% ethanol (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0651
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.