Learn More
Abcam Ag 825, ErbB2 Inhibitor, Solid, 10 mg
Description
- Molecular Weight: 397.5 Da
- Purity: >96%
- Form: Solid
- Activity: Selective ErbB2 inhibitor (IC50 values are 0.15 μM at ErbB2 and 19 μM at ErbB1)
- Biological effects: Suppresses IL-6 induced STAT3 activation, shows antiproliferative effects, and shows antitumor effects in vivo
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | ErbB2, ErbB1 |
| Shipping Condition | Ambient |
| Alternate Part Number | 149092-50-2 |
| Buffer | 100% AG 825 |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% AG 825 |
| Quantity | 10 mg |
| For Use With (Application) | ErbB2 inhibition, STAT3 activation suppression, antiproliferative research, antitumor research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.