Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Ag 825, ErbB2 Inhibitor, Solid, 50 mg

Catalog No. 502705139
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5139 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5139 Supplier Abcam Supplier No. AB14556650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective ErbB2 inhibitor that suppresses IL-6 induced STAT3 activation and exhibits antiproliferative and antitumor effects.

  • Selective inhibitor: ErbB2 (IC50 values are 0.15 μM at ErbB2 and 19 μM at ErbB1).
  • Biological activity: Suppresses IL-6 induced STAT3 activation, and shows antiproliferative and antitumor effects in vivo.
  • Purity: >96%.
  • Molecular weight: 397.5 Da.
  • Form: Solid.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule ErbB2, ErbB1
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Content And Storage Store at -20°C.
Form Solid
Alternate Part Number 149092-50-2
Buffer 100% AG 825
Quantity 50 mg
For Use With (Application) ErbB2 inhibition, STAT3 pathway suppression, antiproliferative research, antitumor research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.