Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521741 Supplier Invitrogen™ Supplier No. PA521741
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, C8D30, NIH-3T3, C2C12, HeLa, Rat2. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GTPase-activating protein for ARF1 and, to a lesser extent, ARF5. Directly and specifically regulates the adapter protein 3 (AP-3)-dependent trafficking of proteins in the endosomal-lysosomal system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGAP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 10% glycerol and 0.01% thimerosal; pH 7
Gene AGAP1
Gene Accession No. Q8BXK8, Q9UPQ3
Gene Alias AGAP1; AGAP-1; Arf GAP with GTP-binding protein-like, ANK repeat and PH domains 1; ArfGAP with GTPase domain, ankyrin repeat and PH domain 1; arf-GAP with GTPase, ANK repeat and PH domain-containing protein 1; centaurin, gamma 2; centaurin-gamma-2; CENTG2; cnt-g2; GGAP1; GTP-binding and GTPase-activating protein 1; Kiaa1099; mKIAA1099; R75364
Gene Symbols AGAP1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human AGAP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116987, 316611, 347722
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.