Learn More
Fisher Science Education™ Agar Bacteriological
SCI_ED
Catalog No. S25127 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
S25127 100 g
1 options
Catalog No. S25127
Add to Cart
Add to Cart
Less hazardous for usual laboratory handling.
  • Non-flammable
  • Insoluble in cold water, but soluble in boiling water forming gels
This product is an Educational Grade Chemical — no C of A is available.
Specifications
Form Solid
Product Type Agar
Quantity 100 g