Learn More
Bioworld Agar, High Gel Strength, 5 kg
Supplier:  Bioworld 401000644
Encompass_Preferred
AgarPurified high gel strength agar with excellent properties for tissue culture and microbiological workGel Strength: 900 g/cm2Working Concentration: 5.0 g/LSulfated ash: <6%This product is sold for laboratory research use only.HS Code : 130231000RTECS Number: AW7950000
Catalog No. 50-255-208
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.