Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Agar, NF Grade

Catalog No. MP21002691 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21002691 1 kg
ICN10026283 250 g
ICN10026280 100 g
ICN10026290 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP21002691 Supplier MP Biomedicals™ Supplier No. 0210026291
Add to Cart
Add to Cart

Powder, meets N.F. grade

  • 80-100 mesh
  • Substitute for gelatin, isinglass
  • Used as nutrient media for bacterial culture
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Form Powder
Grade USP, NF
Product Type Nutrient Agar
For Use With (Application) Used for nutrient media for bacterial cultures
Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.