Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Agar Powder
GREENER_CHOICESCI_ED

Catalog No. S81678 Shop All Supertek Scientific Products
Change view
Click to view available options
Quantity:
0.5 L (Foil Bag)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S81678 0.5 L (Foil Bag)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81678 Supplier Supertek™ Supplier No. CH14032S3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nutrient Agar is used as a culture medium to cultivate various household bacteria

  • This bag of dry nutrient agar will make 500 mL of agar (1/2 liter)
  • Detailed agar preparation instructions included
  • This is enough agar for twenty 90 mm petri dishes
  • Age 7+

Specifications

Appearance Beige Powder
Biosafety Level 0
Content And Storage Keep Dry
Form Powder
Manufacturing Origin United States
Media Type Nutrient Agar
Product Type Tubed Media
Sterility Sterile
Target Organism Class Bacteria
Test Type Standard Plate Count for Microorganism
Fill Volume 500 mL
For Use With (Application) Culture Medium
Quantity 0.5 L (Foil Bag)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.