Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™ PROMO

Catalog No. BP135650S24 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP135650S24 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP135650S24

Synonym: Agarotetraose, Sepharose, Agarose LMP

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C24H38O19
Molecular Weight (g/mol) 630.55
InChI Key MJQHZNBUODTQTK-WKGBVCLCSA-N
IUPAC Name (2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Physical Form Powder/Solid
Color Cream
Quantity 500 g
DNase Dnase-free
Electroendosmosis (EEO) 0.15 Maximum
Gelation Temperature 34.5° to 37.5°C
Gel Strength 1200 g/cm2min.
Packaging Poly Bottle
Formula Weight 306.12
Grade DNase- and RNase-free
Chemical Name or Material Agarose, Broad Separation Range for DNA/RNA (500 bp to 25 kb)
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.