Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Agarose IEF Media

Catalog No. 45000080 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-080 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-080 Supplier Cytiva Supplier No. 17046801
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use for isoelectric focusing of large molecules. Agarose IEF Media is the choice for many IEF applications and immunochemical detection methods.

  • Gelling temp 40 to 43°C
  • Gel strength (Kobe) 1.5%, >1000

TRUSTED_SUSTAINABILITY

Specifications

Color White
Separation Type Isoelectric Focusing
Form Dried
For Use With (Application) Protein purification
Melting Temperature 60° to 90°C
Quantity 10 g
Storage Requirements Dry at room temperature
Product Type Agarose IEF Media
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.