Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Agarose low EEO, for electrophoresis, (Mr <= 0.12)

Catalog No. AC159820250
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159820250 25 g Glass Bottle
AC159820100 10 g Glass Bottle
AC159821000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC159820250 Supplier Thermo Scientific Chemicals Supplier No. 159820250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 9012-36-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Fine Crystalline Powder
Color White
Quantity 25 g
Electroendosmosis (EEO) electroendosmosis (-Mr): 0.07 to 0.12
Gelation Temperature 40 to 43°C (1.5% sol.)
Gel Strength >1800g/cm2 (1.5% gel)
Infrared Spectrum Authentic
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in hot water. Other solubilities: insoluble in most common organic solvents
Grade Electrophoresis
Chemical Name or Material Agarose low EEO
Show More Show Less
EINECSNumber 232-731-8
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.