Learn More
AG Scientific Inc Agarose, Low Melt, 50 G
Supplier:  AG Scientific Inc A273350GM
Encompass_Preferred
Ideal gelling properties for use in cloning assays or for recovery of heat labile samples following electrophoresis.CAS Number: 9012-36-6Solubility: WaterStorage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7081
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.