Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agarose, Molecular Biology Grade, Hoefer™
SDP

Catalog No. HBGR14025
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
2 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HBGR14025 25 g
03-500-523 500 g
GR140500X 2 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. HBGR14025 Supplier Hoefer, Inc Supplier No. GR14025
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9012-36-6

Specifications

Physical Form Granular Free-flowing Powder
Quantity 25 g
Grade Molecular Biology
Chemical Name or Material Agarose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.