Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agarose Multipurpose, MP Biomedicals™

Catalog No. MP11AGP0100 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
3 x 100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP11AGP0100 100 g
MP11AGP0300 3 x 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP11AGP0100 Supplier MP Biomedicals, Inc Supplier No. 11AGAP0100
Add to Cart
Add to Cart

Agarose Multipurpose, Molecular Biology Grade, White powder

  • Agarose is a natural polysaccharide isolated from agar or agar-bearing marine algae.
  • Its physical and chemical properties are particularly suited for use as a gel medium for diffusion and electrokinetic movement of biomolecules.
  • Agarose is essentially biologically inert and forms relatively clear, transparent gels.
  • Agarose based gels offer several advantages: High gel strength permits use of low concentrations.
  • The macroporous nature of the gels allows rapid diffusion of high molecular weight molecules. Agarose is Non-toxic.
  • Agarose gels are thermally reversible facilitating sample recovery.
  • Staining and destaining may be done rapidly with minimal background.
  • Because agarose forms macroporous gels that are electrically nonionic, it is an excellent medium for Isoelectric Focusing (IEF).
  • A selection of different formulated agaroses with specific qualities for unique applications are available from MP.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C24H38O19
Molecular Weight (g/mol) 630.55
MDL Number MFCD00081294
InChI Key MJQHZNBUODTQTK-WKGBVCLCSA-N

Specifications

Physical Form Powder
Melting Point 89°C ±1.5°C (1.5% gel)
Color White
pH 8.21 (in solution), 7.98 (in gel)
Quantity 100 g
Electroendosmosis (EEO) < 0.12%
Gelation Temperature 36 ± 1.5°C, 1.5%gel
Gel Strength ≥3200g/cm2 (1.5%gel); ≥1,880g/cm2 (1%gel)
Synonym Galactan Hydrocolloid, 3,6-Anhydro-α-L-galacto-β-D-galactan, FastLane agarose, Indubiose A4
Solubility Information Soluble in water (1.5g plus 100 mL).
Grade Molecular Biology
Show More Show Less
RTECSNumber AW795000
storageNote1 Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.