Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGFG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530234 Supplier Invitrogen™ Supplier No. PA530234
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: IMR32. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (88%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

HRB is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. HRB binds the activation domain of the human immunodeficiency virus Rev protein when Rev is assembled onto its RNA target, and is required for the nuclear export of Rev-directed RNAs. The protein encoded by this gene is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. This encoded protein binds the Rev activation domain when Rev is assembled onto its RNA target and can significantly enhance Rev activity when overexpressed. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGFG1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene AGFG1
Gene Accession No. P52594, Q8K2K6
Gene Alias 0710001B06Rik; AGFG1; Agfg1-ps1; AI838505; arf-GAP domain and FG repeat-containing protein 1; arf-GAP domain and FG repeats-containing protein 1; arf-GAP domain and FG repeats-containing protein 1; arf-GAP domain and FG repeat-containing protein 1; ArfGAP with FG repeats 1; AU045498; C130049H11Rik; C85612; D730048C23Rik; HIV-1 Rev binding protein; HIV-1 Rev-binding protein; HIV-1 Rev-binding protein homolog; Hrb; hRIP, Rev interacting protein; MGC116938; MGC116940; NEUG(55-78); Neurogranin; neurogranin (protein kinase C substrate, RC3); Ng; NG/RC3; Nrgn; Nucleoporin-like protein RIP; postsynaptic spine 1; protein kinase C substrate; Pss1; R75334; RAB; Rab, Rev/Rex activation domain-binding protein; RC3; rev/Rex activation domain-binding protein; Rev-interacting protein; Rip; Unknown (protein for MGC:128795)
Gene Symbols AGFG1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RIP/Rab. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15463, 3267
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.