Learn More
Morwell MD Biosciences AGGRECAN MONO ANTIBDY 100UG
SDP
Supplier:  Morwell MD Biosciences 1042003
Encompass
Aggrecan monoclonal antibody to C-terminal neoepitope NITEGE /mouse clone BC-13/. This aggrecan degradation product usually remains within the tissue still complexed to hyaluronan and link protein. Its release from the cartilage usually signals that there has been extensive catabolism of aggrecan which allows large complexes containing this metabolite to be released from the tissue.
Catalog No. NC3862893