Learn More
ATCC Aggregatibacter; actinomycetemcomitans
Supplier:  ATCC 43718
Encompass
This bacterial strain was isolated from subgingival dental plaque. It grows in a 5% CO2 atmosphere on brain heart infusion medium.
Catalog No. NC2313905
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.