Learn More
Abcam Aggresome Detection Kit, Cell-based, Fluorescent, 100/Test
Description
- Assay type: Cell-based
- Detection method: Fluorescent
- Sample types: Suspension cells, Adherent cells
- Applications: Flow cytometry, Fluorescence microscopy
- Citations: Cited in 11 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Aggresome |
| No. of Reactions | 100 Tests |
| For Use With (Equipment) | Flow Cytometer, Fluorescence Microscope |
| Detection Method | Fluorescence |
| Format | Kit |
| Detection Location | In-Cell Detection |
| Sample Type | Suspension cells, Adherent cells |
| Quantity | 100 Tests |
| For Use With (Application) | Aggresome detection, Flow cytometry, Fluorescence microscopy |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.