Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGK 2, Tocris Bioscience™

Catalog No. 323350
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-335-0 50 mg
32-331-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-335-0 Supplier R&D Systems Supplier No. 3233/50
This item is not returnable. View return policy

Selective SIRT2 inhibitor

Selective inhibitor of SIRT2 (IC50 = 3.5 μM). Displays no activity at SIRT1 and SIRT3 at concentrations up to 40 μM. Reduces α-synuclein-mediated toxicity in in vitro and in vivo models of Parkinson's disease.

Chemical Identifiers

CAS 304896-28-4
Molecular Formula C23H13Cl2N3O2
Molecular Weight (g/mol) 434.28
MDL Number MFCD01909444
InChI Key SVENPFFEMUOOGK-UHFFFAOYSA-N
PubChem CID 2130404
IUPAC Name 2-cyano-3-[5-(2,5-dichlorophenyl)furan-2-yl]-N-(quinolin-5-yl)prop-2-enamide
SMILES ClC1=CC(C2=CC=C(O2)C=C(C#N)C(=O)NC2=C3C=CC=NC3=CC=C2)=C(Cl)C=C1

Specifications

Quantity 50 mg
Synonym agk2, agk-2, agk 2, unii-ddf0l8606a, sirtuin 2 inhibitor, sirt2 inhibitor, agk2, 2-cyano-3-5-2,5-dichlorophenyl-2-furanyl-n-5-quinolinyl-2-propenamide, e-2-cyano-3-5-2,5-dichlorophenyl furan-2-yl-n-quinolin-5-ylprop-2-enamide, 2-cyano-3-5-2,5-dichlorophenyl furan-2-yl-n-quinolin-5-yl acrylamide
Formula Weight 434.27
Percent Purity >98%
Chemical Name or Material AGK 2
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.