Learn More
Apexbio Technology LLC AGK7 304896-21-7 1mg
SDP
Supplier:  Apexbio Technology LLC C57091
Encompass_Preferred
AGK7 (CAS 304896-21-7) is a small molecule used as an inactive control compound for AGK2 a selective inhibitor of the NAD -dependent deacetylase SIRT2 While AGK2 potently inhibits SIRT2 activity (IC50 3 5 M) and increases cellular acetylated tubulin levels AGK7 lacks this inhibitory effect Notably in cellular models AGK2 but not AGK7 reduces -synuclein-mediated toxicity and modulates protein aggregation AGK7 serves as a specificity control in studies investigating the role of SIRT2 in cellular processes such as cell cycle regulation neurodegeneration and epigenetic modification
Catalog No. 50-000-92788
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.