Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Agk7, Inactive Control, Solid, >99% Purity, 5 mg

Catalog No. 502705639
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5639 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5639 Supplier Abcam Supplier No. AB1445485MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-permeable inactive control and structural isomer of the SIRT2 inhibitor AGK2, with >99% purity.

  • Molecular Weight: 434.3 Da
  • Purity: >99%
  • Form: Solid
  • Properties: Inactive control; structural isomer of AGK2 (ab142073) (SIRT2 inhibitor). Cell-permeable.
  • Storage: Store at Ambient.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Content And Storage Store at ambient temperature.
Form Solid
Quantity 5 mg
For Use With (Application) Inactive control
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.