Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGO2 Recombinant Rabbit Monoclonal Antibody (036)
GREENER_CHOICE

Catalog No. PIMA529649
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529649 Supplier Invitrogen™ Supplier No. MA529649
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse AGO2.

Ago proteins are broadly expressed in somatic cells, associate with miRNAs and are key actors in different RNA silencing pathways. However, only the Ago2 protein displays endonucleolytic or Slicer activity and can therefore execute miRNA-directed cleavage of target mRNA, provided that the base-pairing between the Ago2-associated miRNA and the mRNA sequence is perfect. In case of partial complementarity, the Ago2 protein fails to cleave, but instead interferes with translation of the target mRNA via its translational repression activity. Ago2 has been shown to play a non-redundant role in small RNA guided gene silencing processes, including RNA interference, translation repression and heterochromatinization. As a core element of the RISC complex, AGO2 initiates the degradation of target mRNAs through its catalytic activity in gene silencing processes guided by siRNAs or miRNAs. In addition to mRNA degradation or gene silencing guided by miRNAs, Ago2 also acts as a RNA slicer in a Dicer-independent way, as well as a regulator of miRNA maturation. Over-expression of Ago2 has been correlated with several aspects of cancers, including tumor cell growth and the overall survival of cancer patients. Furthermore, gene disruption in the mouse demonstrated that the Ago2 protein is essential for embryonic development and a key regulator of B-lymphoid and erythroid development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGO2
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Recombinant Monoclonal
Clone 036
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene AGO2
Gene Accession No. Q8CJG0
Gene Alias 1110029L17Rik; 2310051F07Rik; AGO1; AGO2; AI225898; AL022874; argonaute 2; argonaute 2, RISC catalytic component; argonaute RISC catalytic component 2; argonaute RISC catalytic subunit 2; Argonaute1; argonaute2; argonaute2 {ECO:0000255; AW546247; CTA-204B4.6; eIF2C; eIF2C 1; eIF-2C 1; eIF2C 2; eIF-2C 2; eIF2C 2 {ECO:0000255; eIF-2C 2 {ECO:0000255; EIF2C1; Eif2c2; ENSMUSG00000072493; Eukaryotic translation initiation factor 2C 2; eukaryotic translation initiation factor 2C 2 {ECO:0000255; eukaryotic translation initiation factor 2C, 2; GERp95; Gm10365; Golgi ER protein 95 kDa; hAgo1; hAgo2; HAMAP-Rule:MF_03031}; Kiaa4215; mAgo2; mKIAA4215; PAZ Piwi domain protein; Piwi/Argonaute family protein meIF2C2; PPD; Protein argonaute-1; protein argonaute-2; Protein slicer; Q10
Gene Symbols AGO2
Host Species Rabbit
Immunogen Recombinant Mouse AGO2 protein (Met1-Ala860).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239528
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.