Learn More
CPC Scientific Ac-Cys-Val-Arg-Leu-His-Glu-Ser-Cys-Leu-Gly-Gln-Gln-Val-Pro-Cys-Cys-Asp-Pro-Cys-Ala-Thr-Cys-Tyr-Cys-Arg-Phe-Phe-Asn-Ala-Phe-Cys-Tyr-Cys-Arg-Lys-Leu-Gly-Thr-Ala-Met-Asn-Pro-Cys-Ser-Arg-Thr-OH (trifluoroacetate salt) 0.1MG
SDP
Supplier:  CPC Scientific AGOU002A
Encompass
ONE-LETTER SEQUENCE: Ac-CVRLHESCLGQQVPCCDPCATCYCRFFNAFCYCRKLGTAMNPCSRT (Cys1 and 16 bridge, Cys8 and 22 bridge, Cys15 and 33 bridge, Cys19 and 43 bridge, Cys24 and 31 bridge)MOLECULAR FORMULA: C219H339N65O63S11MOLECULAR WEIGHT:5233.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Obesity
Catalog No. 50-210-8642
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.