Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGPAT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101343
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101343 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101343 Supplier Invitrogen™ Supplier No. PA5101343
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AGPAT3.

The protein encoded by this gene is an acyltransferase that converts lysophosphatidic acid into phosphatidic acid, which is the second step in the de novo phospholipid biosynthetic pathway. The encoded protein may be an integral membrane protein. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGPAT3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Agpat3
Gene Accession No. Q9D517, Q9NRZ7
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 3; 1-acyl-sn-glycerol-3-phosphate acyltransferase gamma; 1-AGP acyltransferase 3; 1-AGPAT 3; Agpat3; AW061257; AW493985; D10Jhu12e; lpaat3; LPAAT-gamma; LPAAT-GAMMA1; lysophosphatidic acid acyltransferase 3; lysophosphatidic acid acyltransferase gamma; lysophosphatidic acid acyltransferase-gamma1; UNQ759/PRO1490
Gene Symbols Agpat3
Host Species Rabbit
Immunogen A synthesized peptide derived from human AGPAT3(Accession Q9NRZ7), corresponding to amino acid residues E125-C175.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28169, 294324, 56894
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.