Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGPAT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549623
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549623 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549623 Supplier Thermo Scientific Supplier No. PA549623
Only null left
Add to Cart
Add to Cart

AGPAT3 Polyclonal Antibody

AGPAT3 Polyclonal Antibody for Western Blot, IHC (P), Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen AGPAT3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Agpat3
Gene Accession No. Q9NRZ7
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 3; 1-acyl-sn-glycerol-3-phosphate acyltransferase gamma; 1-AGP acyltransferase 3; 1-AGPAT 3; Agpat3; AW061257; AW493985; D10Jhu12e; lpaat3; LPAAT-gamma; LPAAT-GAMMA1; lysophosphatidic acid acyltransferase 3; lysophosphatidic acid acyltransferase gamma; lysophosphatidic acid acyltransferase-gamma1; UNQ759/PRO1490
Gene Symbols Agpat3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 241-269 amino acids from the Central region of human AGPAT3.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56894
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.