Learn More
ATCC Agrobacterium tumefaciens Strain A 1 (ATCC 4720) - Type strain, freeze-dried research culture
Supplier:  ATCC 4720
Encompass
Agrobacterium tumefaciens strain A 1 (ATCC 4720) is the authenticated type strain supplied for laboratory research. This whole-genome sequenced bacterial culture is provided freeze-dried and intended for research use only; it is not suitable for therapeutic, diagnostic, or consumption purposes.
  • Type strain; whole-genome sequenced.
  • Supplied freeze-dried; store at 2°C to 8°C.
  • Grow on nutrient agar or nutrient broth at 26°C under aerobic conditions.
  • Biosafety level 1 organism.
  • For laboratory research use only; not for therapeutic, diagnostic, or consumption use.
  • USDA permit required for movement of live plant pests, noxious weeds, and soil; Hawaii import permit documentation may be required for shipments to Hawaii.
Catalog No. NC2885988