Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ AGTR-1 Antibody Blocking Peptide
SDP

Catalog No. NB814339 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB814339 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB814339 Supplier Novus Biologicals™ Supplier No. NB10057073PEP
Add to Cart
Add to Cart

Specifications

Gene ID (Entrez) 185
Species Human
Purity >50%, by HPLC
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-57073
Gene Alias AG2S, AGTR1AAngiotensin II type-1 receptor, Angiostensin Receptor, angiotensin II receptor, type 1, angiotensin receptor 1, angiotensin receptor 1 B, AT1AT1AR, AT1B, AT1R, AT2R1A, AT2R1AGTR1B, AT2R1BAT1BR, HAT1R, type-1 angiotensin II receptor, type-1 B angiotensin II receptor
Gene Symbol AGTR1
Quantity 0.1 mg
Research Category GPCR
Storage Buffer Lyophilized from a 5 pmol/ul solution in 5% (v/v) acetonitrile/H2O and TFA salt.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.