Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGTR1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2C5G11, Proteintech
SDP

Catalog No. 501736842 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6842 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6842 Supplier Proteintech Group Inc Supplier No. 664151IG150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

AGTR1 is known as Angiotensin II Type 1 Receptor. Angiotensin II is a potent vasopressor hormone and a primary regulator of aldosterone secretion. It is an important effector controlling blood pressure and volume in the cardiovascular system. And it acts through at least two types of receptors. This gene encodes the type 1 receptor which is thought to mediate the major cardiovascular effects of angiotensin II. Moreover, this gene may play a role in the generation of reperfusion arrhythmias following restoration of blood flow to ischemic or infarcted myocardium. It was previously thought that a related gene, denoted as AGTR1B, existed; however, it is now believed that there is only one type 1 receptor gene in humans. Multiple alternatively spliced transcript variants have been reported for this gene.

Specifications

Antigen AGTR1
Applications Western Blot
Classification Monoclonal
Clone 2C5G11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AGTR1
Gene Accession No. P25095, P29754, P30555, P30556
Gene Alias AG2S, AGTR1, AGTR1A, AGTR1B, Angiotensin II type 1 receptor, AT1, AT1AR, AT1B, AT1BR, AT1R, AT2R1, AT2R1A, AT2R1B, HAT1R, Type 1 angiotensin II receptor
Gene Symbols AGTR1, Agtr1a
Host Species Mouse
Immunogen AGTR1 Fusion Protein Ag14461
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100519976, 11607, 185, 24180
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.