Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGTR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106837 Supplier Invitrogen™ Supplier No. PA5106837
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AGTR1.

AGTR1 is known as Angiotensin II Type 1 Receptor. Angiotensin II is a potent vasopressor hormone and a primary regulator of aldosterone secretion. It is an important effector controlling blood pressure and volume in the cardiovascular system. And it acts through at least two types of receptors. This gene encodes the type 1 receptor which is thought to mediate the major cardiovascular effects of angiotensin II. Moreover, this gene may play a role in the generation of reperfusion arrhythmias following restoration of blood flow to ischemic or infarcted myocardium. It was previously thought that a related gene, denoted as AGTR1B, existed; however, it is now believed that there is only one type 1 receptor gene in humans. Multiple alternatively spliced transcript variants have been reported for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGTR1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AGTR1
Gene Accession No. P25095, P29754, P30556
Gene Alias 1810074K20Rik; AG2S; AGTR1; Agtr1a; Agtr-1a; AGTR1B; AI551199; angiotensin II receptor type 1; angiotensin II receptor, type 1; angiotensin II receptor, type 1a; Angiotensin II type-1 receptor; Angiotensin II type-1A receptor; angiotensin receptor 1; angiotensin receptor 1a; Angtr-1a; AT1; At1a; AT1AR; AT1B; AT1BR; AT1R; AT2R1; AT2R1A; AT2R1B; HAT1R; Type-1 angiotensin II receptor; Type-1A angiotensin II receptor; type-1B angiotensin II receptor; vascular type-1 angiotensin II receptor
Gene Symbols AGTR1, Agtr1a
Host Species Rabbit
Immunogen A synthesized peptide derived from human AGTR1(Accession P30556), corresponding to amino acid residues M1-L34.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11607, 185, 24180
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.