Learn More
Selleck Chemical LLC AH 6809-E0648-5MG
SDP
Supplier:  Selleck Chemical LLC E06485MG
Encompass_Preferred
AH 6809 is an antagonist of EP and DP receptor with Kis of 1217 1150 1597 and 1415 nM for the cloned human EP1 EP2 EP3-III and DP receptor respectively AH 6809 has a Ki of 350 nM for mouse EP2 receptor
Catalog No. 50-313-7732
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.