Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AHA1 Monoclonal Antibody (25F2.D10), APC
GREENER_CHOICE

Catalog No. PIMA545237
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545237 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545237 Supplier Invitrogen™ Supplier No. MA545237
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

1 μg/mL of MA5-45237 was sufficient for detection of Aha1 in 10 μg of rat tissue lysate by colorimetric immunoblot analysis using Goat anti-rat IgG:HRP as the secondary antibody. Detects approximately 38kDa. Can run up to 45kDa on SDS Page.

Acts as a co-chaperone of HSP90AA1 (PubMed:29127155). Activates the ATPase activity of HSP90AA1 leading to increase in its chaperone activity (PubMed:29127155). Competes with the inhibitory co-chaperone FNIP1 for binding to HSP90AA1, thereby providing a reciprocal regulatory mechanism for chaperoning of client proteins (PubMed:27353360). Competes with the inhibitory co-chaperone TSC1 for binding to HSP90AA1, thereby providing a reciprocal regulatory mechanism for chaperoning of client proteins (PubMed:29127155). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AHA1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 25F2.D10
Concentration 1.59 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.6
Gene Ahsa1
Gene Accession No. O95433, Q8BK64
Gene Alias Activator of 90 kDa heat shock protein ATPase homolog 1; activator of Hsp90 ATPase activity 1; Activator of Hsp90 ATPase protein 1; Aha 1; AHA1; AHA1, activator of heat shock 90kDa protein ATPase homolog 1; AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast); AHA1, activator of heat shock protein ATPase 1; AHA1, activator of heat shock protein ATPase homolog 1; Aha1p; AHSA 1; Ahsa1; BC023857; C14orf3; hAha1; Hsp90 co-chaperone AHA1; HSPC322; p38; SHSA1; YD8142.16; YD8142B.06; YDR214W
Gene Symbols Ahsa1
Host Species Rat
Immunogen Mouse Aha1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10598, 217737, 681996
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.