Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AHNAK Monoclonal Antibody (EM-09)
GREENER_CHOICE

Catalog No. PIMA110050 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA110050 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA110050 Supplier Invitrogen™ Supplier No. MA110050
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This antibody reacts with AHNAK1, a 700 kDa multi-functional adaptor protein expressed mainly in epithelial cell, various types of muscle cells and immune cells. Immunocytochemistry: permeabilization is required.

AHNAK1 (Desmoyokin) is a large (700 kDa) scaffold protein that translocates to the plasma membrane after an increas of extracellular calcium level or upon proteinkinase C activation and regulates extracellular calcium influx mediated by L-type Ca2+ channels. AHNAK1 has been implicated in diverse signal transduction proceses affecting cell differentiation and proliferation. In response to calcium-dependent intercellular contacts AHNAK1 forms multimeric complexes in the plasma membrane, connected with actin and annexin 2/S100A10 assemblies and is thus involved in organization of the plasma membrane architecture. In epithelial cells, AHNAK1 is localized in cytoplasm or is membrane-associated, but in cells of nonepithelial origin AHNAK1 is predominantly nuclear; it has a weak DNA-binding activity and associates with the DNA-ligase IV-XRCC4 complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AHNAK
Applications Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone EM-09
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene AHNAK
Gene Accession No. Q09666
Gene Alias AHNAK; AHNAK 1; AHNAK nucleoprotein; AHNAK nucleoprotein (desmoyokin); AHNAK-related; AHNAKRS; Desmoyokin; DY6; MGC5395; Neuroblast differentiation-associated protein AHNAK; PM227; RGD1308572; UV118
Gene Symbols AHNAK
Host Species Mouse
Immunogen Bacterially expressed fragment of N-terminal domain of human AHNAK1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66395, 79026
Target Species Human, Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.