Learn More
Medchemexpress LLC Aica-riboside, 5'-phosphate | 3031-94-5 | MFCD00057264 | 100.0% | C9H15N4O8P | 10MG
Supplier: Medchemexpress LLC HY13435610MG
AICA-riboside, 5′-phosphate is the 5′-phosphorylated form of AICA-riboside that functions as an AMP mimetic and activator of AMP-activated protein kinase (AMPK). It is provided as a high-purity solid for biochemical and cellular research into metabolic and neurological pathways.
- Acts as an AMP mimetic and activates AMP-activated protein kinase.
- Provided as a high-purity solid suitable for biochemical studies.
- Molecular formula C9H15N4O8P and molecular weight 338.21 g·mol⁻¹.
- Available in small quantities for experimental use.
- Commonly used in metabolic disease and neuroprotection research.